Anti-CEACAM3 Rabbit polyclonal antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: CEACAM3 (carcinoembryonic antigen-related cell adhesion molecule 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- BIRBORB135264-100
- BIRBORB135264-200
- BIRBORB135264-50
- Antigen symbol:
- CEACAM3
- CEACAM3
- CEACAM3
- Antigen name:
- Carcinoembryonic antigen-related cell adhesion molecule 3
- Carcinoembryonic antigen-related cell adhesion molecule 3
- Carcinoembryonic antigen-related cell adhesion molecule 3
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Storage temperature:
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice