Anti-MAP7 Rabbit Polyclonal Antibody
Catalog # BIRBORB131795-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:microtubule-associated Protein 7
- Antigen symbol:MAP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage buffer:1 x PBS (pH 7,4), 40% Glycerol
- Molecular weight:84 kDa
- Storage temperature:Cool
- Concentration:100 ug/ml BSA
- Shipping temperature:Ice
- Immunogen:N-terminal or the C-terminal region of human MAP7
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:1000-1:3000
Type: Primary
Antigen: MAP7 (microtubule-associated protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: