Anti-SUPT16H Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: SUPT16H (suppressor of Ty 16 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- BIRBORB127254-200
- BIRBORB127254-100
- Antigen symbol:
- SUPT16H
- SUPT16H
- Antigen name:
- suppressor of Ty 16 homolog (S. cerevisiae)
- suppressor of Ty 16 homolog (S. cerevisiae)
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice