Anti-ADNP Rabbit Polyclonal Antibody
: Biorbyt
Type: Primary
Antigen: ADNP (activity-dependent neuroprotector homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
- Catalog No:
- BIRBORB126711-100
- BIRBORB126711-200
- Antigen symbol:
- ADNP
- ADNP
- Antigen name:
- activity-dependent neuroprotector homeobox
- activity-dependent neuroprotector homeobox
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice