Anti-AIFM1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
BIRBORB124441-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Apoptosis-inducing factor, mitochondrion-associated 1
- Antigen symbol:AIFM1
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:57 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: AIFM1 (apoptosis-inducing factor, mitochondrion-associated, 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: