Anti-IFNAR1 Rabbit Polyclonal Antibody (Cy3®)
BIRBORB114663-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Interferon (alpha, beta and omega) receptor 1
- Antigen symbol:IFNAR1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Modification:phospho-Ser535, phospho-Ser539
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:90 - 130 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human IFNAR1 around the phosphorylation site of Ser535+Ser539.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: IFNAR1 (interferon (alpha, beta and omega) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat