Anti-CXCR4 Rabbit Polyclonal Antibody (Biotin)
Catalog # BIRBORB114011-100
Leverantör: Biorbyt
Specifikationer
- Antibody type:Primary
- Antigen name:chemokine (C-X-C motif) receptor 4
- Antigen symbol:CXCR4
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:39 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: