Anti-CAV1 Rabbit Polyclonal Antibody
BIRBORB109339-50
: Biorbyt
- Antibody type:Primary
- Antigen name:caveolin 1, caveolae Protein, 22kDa
- Antigen symbol:CAV1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:N-terminal region of human
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human