Anti-XIAP Rabbit Polyclonal Antibody
BIRBORB108955-100
: Biorbyt
- Antibody type:Primary
- Antigen name:X-linked inhibitor of apoptosis
- Antigen symbol:XIAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Rabbit polyclonal AIM antibody was raised against a 13 amino acid peptide from near the amino terminus of human AIM.
- Purification:purified by antigen immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: