Anti-G 5-D1 Rabbit polyclonal antibody
BIRBORB108893-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:G 5-D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Unique peptide sequence derived from the D1 blade of the G 5 protein.
- Purification:purified by ammonium sulfate precipitation
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: G 5-D1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: