Anti-Lp-PLA2 None Antibody
Catalog # BIRBORB108119-0.5
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:LP-PLA2
- Conjugation:Unconjugated
- Isotype:IgG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type: Primary
Antigen: Lp-PLA2
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG
Reactivity: