Anti-PI3K p85 Rabbit Polyclonal Antibody
BIRBORB106105-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:PI3K p85
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Tyr458
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:85 kDa
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human PI3K around the phosphorylation site of p85 Tyr458.
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: PI3K p85
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat