Anti-CDC42BPA Rabbit Polyclonal Antibody
BIRBORB44765-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:CDC42 binding protein kinase alpha (DMPK-like)
- Antigen symbol:CDC42BPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: CDC42BPA (CDC42 binding protein kinase alpha (DMPK-like))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human