Anti-CSDE1 Rabbit Polyclonal Antibody
BIRBORB41917-100
: Biorbyt
- Antibody type:Primary
- Antigen name:cold shock domain containing E1, RNA-binding
- Antigen symbol:CSDE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Amino acid number:167 to 372
- Storage buffer:0,1M Tris, 0,1M Glycine, 20% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:0,77 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 167 and 372 of CSDE1.
- Purification:purified by antigen affinity chromatography
- Size:100 µl
- Pk:100 µl
Western Blot: 1:500-1:3000
Type: Primary
Antigen: CSDE1 (cold shock domain containing E1, RNA-binding)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human