Anti-CACNA1B Rabbit Polyclonal Antibody
Catalog # BIRBORB40926-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, N type, alpha 1B subunit
- Antigen symbol:CACNA1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Amino acid number:2013 to 2179
- Storage buffer:1XPBS, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:0,43 mg/ml
- Shipping temperature:Ice
- Immunogen:2013 - 2179 of Human CACNA1B.
- Purification:purified by antigen affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CACNA1B (calcium channel, voltage-dependent, N type, alpha 1B subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human