Anti-CapZ beta Rabbit Polyclonal Antibody
BIRBORB40854-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:CapZ beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Amino acid number:1 to 266
- Storage buffer:1XPBS, 40% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:2 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 266 of CapZ-beta .
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Western Blot: 1:500-1:3000
Type: Primary
Antigen: CapZ beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human