Anti-DOCK 180 Rabbit Polyclonal Antibody
Catalog # BIRBORB40763-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:DOCK 180
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:1802 to 1865
- Storage buffer:1XPBS, 1%BSA, 20% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide contain a sequence corresponding to a region within amino acids 1802 and 1865 of DOCK 180 .
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250
Type: Primary
Antigen: DOCK 180
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human