Anti-MAP2K3 Rabbit Polyclonal Antibody
Catalog # BIRBORB27589-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 3
- Antigen symbol:MAP2K3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized
- Amino acid number:320 to 334
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Sequence:MEHPFFTLHKTKKTD
- Storage temperature:Cool
- Concentration:500 ug/ml
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human MAP2K3 (320-334aa MEHPFFTLHKTKKTD), identical to the related mouse and rat sequences.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MAP2K3 (mitogen-activated protein kinase kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat