Anti-Neuron Specific Enolase Mouse Monoclonal Antibody
Catalog # BIRBORB26015-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Neuron Specific Enolase
- Antigen symbol:NSE
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:5,1 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Purified neuron-specific enolase from human brain.
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Immunochemistry: 1:100-1:300
Type: Primary
Antigen: Neuron Specific Enolase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: