Order Entry
Sweden
ContactUsLinkComponent
Anti-RNP Antigen Human Polyclonal Antibody
Anti-RNP Antigen Human Polyclonal Antibody
Catalog # BIRBORB25151-2
Supplier:  Biorbyt
Anti-RNP Antigen Human Polyclonal Antibody
Catalog # BIRBORB25151-2
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RNP Antigen
  • Antigen symbol:
    RNP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Human
  • Form:
    lyophilized
  • Storage buffer:
    0,01 M Phosphate, 0,13 M Sodium Chloride, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by delipidation and defibrination
  • Size:
    2 ml
  • Pk:
    2 mL

Specifications

About this item

Type: Primary
Antigen: RNP Antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Human
Isotype: none
Reactivity: