Order Entry
Sweden
ContactUsLinkComponent
Anti-MAPK1/3 Mouse Monoclonal Antibody [clone: E31R]
Anti-MAPK1/3 Mouse Monoclonal Antibody [clone: E31R]
Catalog # BIRBORB22971-100
Supplier:  Biorbyt
Anti-MAPK1/3 Mouse Monoclonal Antibody [clone: E31R]
Catalog # BIRBORB22971-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 1/3
  • Antigen symbol:
    MAPK1/3
  • Clonality:
    Monoclonal
  • Clone:
    E31R
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage buffer:
    Buffer: 10mM PBS, pH 7,2, 0,05% NaN3
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant full length ERK1 protein
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: monoclonal
Clone: E31R
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: