Order Entry
Sweden
ContactUsLinkComponent
Anti-PDCD1 Mouse Monoclonal Antibody
Anti-PDCD1 Mouse Monoclonal Antibody
Supplier:  Biorbyt
Certificates
Anti-PDCD1 Mouse Monoclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB22895-20
  • BIRBORB22895-5
  • BIRBORB22895-100
  • Antigen symbol:
  • PDCD1
  • PDCD1
  • PDCD1
  • Antigen name:
  • Programmed cell death 1
  • Programmed cell death 1
  • Programmed cell death 1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b
  • IgG2b
  • IgG2b
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Anti-human PDCD1 mAb, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with recombinant human PDCD1 amino acids 21-167 purified from E.coli
  • Anti-human PDCD1 mAb, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with recombinant human PDCD1 amino acids 21-167 purified from E.coli
  • Anti-human PDCD1 mAb, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with recombinant human PDCD1 amino acids 21-167 purified from E.coli
  • Amino acid number:
  • 21 to 167
  • 21 to 167
  • 21 to 167
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information