Order Entry
Sweden
ContactUsLinkComponent
Anti-IgA (secretory component) Rabbit Polyclonal Antibody
Anti-IgA (secretory component) Rabbit Polyclonal Antibody
Catalog # BIRBORB22295-1
Supplier:  Biorbyt
Anti-IgA (secretory component) Rabbit Polyclonal Antibody
Catalog # BIRBORB22295-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgA (secretory component)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Format:
    antiserum
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Highly purified secretory IgA isolated from pooled bovine milk. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Secondary
Antigen: Bovine Secretory IgA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: