Anti-Secretory Component Mouse Monoclonal Antibody
Catalog # BIRBORB21171-0.5
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:Secretory Component
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Format:ascites
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Highly purified secretory component isolated from human milk.
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type: Primary
Antigen: Secretory Component
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human