Anti-L-Glutamic Dehydrogenase Rabbit Polyclonal Antibody
Catalog # BIRBORB20718-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:L-Glutamic Dehydrogenase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Bovine
- Form:lyophilized
- Storage buffer:PBS
- Storage temperature:Cool
- Concentration:1 mg/0,5ml, no foreign proteins added
- Shipping temperature:Ice
- Immunogen:L-glutamic dehydrogenase isolated and purified from bovine liver. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: L-Glutamic Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine