Anti-Niban like protein 1 Rabbit Polyclonal Antibody
BIRBORB14701-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Niban like Protein 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Storage buffer:PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:The antiserum was produced against synthesized non-phosphopeptide derived from human niban like protein 1 around the phosphorylation site of serine 712 (Q -V-S-SP-P).
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:1000 Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: Niban like protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: