Anti-CD3 Mouse Monoclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Specifications
- Catalog No:
- BIRBORB11921-100
- BIRBORB11921-500
- Antigen symbol:
- CD3
- CD3
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Storage buffer:
- Phosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSA
- Phosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSA
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice