Anti-STAT1 Rabbit Polyclonal Antibody
Catalog # BIRBORB11426-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Signal Transducer And Activator of Transcription 1
- Antigen symbol:STAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Modification:phospho-Tyr701
- Storage buffer:PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:The antiserum was produced against synthesized phosphopeptide derived from human Stat1 around the phosphorylation site of tyrosine 701 (T -G-YP-I-K).
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:500-1:1000 Immunochemistry: 1:50-1:100
Type: Primary
Antigen: STAT1 (signal transducer and activator of transcription 1, 91kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat