Anti-CCNE1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB8810-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:cyclin E1
- Antigen symbol:CCNE1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Modification:phospho-Thr395
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:32 kDa
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human phospho-Cyclin E(Thr395).
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat