Anti-NEIL2 Mouse Monoclonal Antibody [clone: 1B7]
ABNOH00252969-M01
: Abnova
- Antibody type:Primary
- Antigen name:Nei-like DNA Glycosylase 2
- Antigen symbol:NEIL2
- Clonality:Monoclonal
- Clone:1B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:252969
- Antigen synonyms:NEH2|NEI2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NEIL2
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human