Anti-ADAL Mouse Polyclonal Antibody
Catalog # ABNOH00161823-B01P
Supplier: Abnova
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:ADAL
- Antigensymbol:ADAL
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Mouse
- Reaktivitet:Human
- Western blot:Yes
- Korsad adsorption:No
- Gen-ID:161823
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:50 µg
- Förp.:50 µg
Specifications
About this item
Type: Primary
Antigen: ADAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human