Anti-C1QTNF2 Mouse Polyclonal Antibody
ABNOH00114898-B01P
: Abnova
- Antibody type:Primary
- Antigen name:C1q And tumor necrosis factor related protein 2
- Antigen symbol:C1QTNF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:114898
- Antigen synonyms:zacrp2|CTRP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: C1QTNF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human