Anti-WBSCR22 Mouse Polyclonal Antibody
ABNOH00114049-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Williams Beuren syndrome chromosome region 22
- Antigen symbol:WBSCR22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:114049
- Antigen synonyms:HASJ4442|MERM1|PP3381|HUSSY-3|WBMT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: WBSCR22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human