Anti-MRRF Mouse Monoclonal Antibody [clone: 1D3]
Catalog # ABNOH00092399-M01
Supplier: Abnova
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Mitochondrial Ribosome ReCycling Factor
- Antigensymbol:MRRF
- Klonalitet:Monoclonal
- Klon:1D3
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Mouse
- ImmunKemi:Yes
- Reaktivitet:Human
- Western blot:Yes
- Korsad adsorption:No
- Gen-ID:92399
- Antigen synonymer:RRF|MTRRF|MRFF
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:100 µg
- Förp.:100 µg
Specifications
About this item
Type: Primary
Antigen: MRRF
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human