Order Entry
Sweden
ContactUsLinkComponent
Anti-MPND Mouse Monoclonal Antibody [clone: 1C12]
Anti-MPND Mouse Monoclonal Antibody [clone: 1C12]
Catalog # ABNOH00084954-M01
Supplier:  Abnova
undefined
Anti-MPND Mouse Monoclonal Antibody [clone: 1C12]
Catalog # ABNOH00084954-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MPN Domain Containing
  • Antigen symbol:
    MPND
  • Clonality:
    Monoclonal
  • Clone:
    1C12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    84954
  • Storage buffer:
    In 1X PBS, pH 7,4
  • Sequence:
    ALLEPGAGVLSIYYLGKKFLGDLQPDGRIMWQETGQTFNSPSAWATHCKKLVNPAKKSGCGWASVKYKGQKLDKYKATWLRLHQLHTPATAADESPAS
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Shipping temperature:
    Ice
  • Immunogen:
    FLJ14981 (NP_116257, 84 a.a. ~ 181 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against a partial recombinant FLJ14981.

Recommended Dilutons: Western Blot (1 to 5 µg/ml).

Type: Primary
Antigen: MPND
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human