Order Entry
Sweden
ContactUsLinkComponent
 
Anti-SLC35A3 Mouse Monoclonal Antibody [clone: 4B6]
Catalog # ABNOH00023443-M01
Supplier:  Abnova
undefined
Anti-SLC35A3 Mouse Monoclonal Antibody [clone: 4B6]
Catalog # ABNOH00023443-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 35 Member A3
  • Antigen symbol:
    Solute carrier family 35, member 3
  • Clonality:
    Monoclonal
  • Clone:
    4B6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    23443
  • Antigen synonyms:
    AMRS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SLC35A3
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human