Anti-PUM2 Mouse Monoclonal Antibody [clone: 3E6]
Catalog # ABNOH00023369-M13
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Pumilio 2
- Antigen symbol:PUM2
- Clonality:Monoclonal
- Clone:3E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23369
- Antigen synonyms:pumilio homolog 2|OTTHUMP00000200539|MGC138253|PUML2|FLJ36528|PUM2|MGC138251|KIAA0235|pumilio (Drosphila) homolog 2|PUMH2|pumilio homolog 2 (Drosophila)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PUM2
Clonality: Monoclonal
Clone: 3E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human