Anti-ZWINT Mouse Monoclonal Antibody [clone: 1B7]
Catalog # ABNOH00011130-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ZW10 interacting kinetochore protein
- Antigen symbol:ZWINT
- Clonality:Monoclonal
- Clone:1B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11130
- Antigen synonyms:ZWINT1|human ZW10 interacting protein-1|zwint-1|KNTC2AP|ZW10-interacting protein 1|ZW10 interactor|HZwint-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ZWINT
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human