Anti-RBM5 Mouse Monoclonal Antibody [clone: 3G6]
Catalog # ABNOH00010181-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:RNA binding motif protein 5
- Antigen symbol:RBM5
- Clonality:Monoclonal
- Clone:3G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10181
- Antigen synonyms:G15|RMB5|H37|LUCA15
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RBM5
Clonality: Monoclonal
Clone: 3G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human