Anti-GTF2IRD1 Mouse Monoclonal Antibody [clone: 2C7]
Catalog # ABNOH00009569-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:GTF2I repeat domain containing 1
- Antigen symbol:GTF2IRD1
- Clonality:Monoclonal
- Clone:2C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9569
- Antigen synonyms:RBAP2|hMusTRD1alpha1|WBSCR11|WBS|CREAM1|GTF3|MUSTRD1|BEN|WBSCR12
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GTF2IRD1
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human