Anti-B4GALT5 Mouse Monoclonal Antibody [clone: 7E6]
ABNOH00009334-M05A
: Abnova
- Antikroppstyp:Primär
- Antikroppsnamn:UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 5
- Antigensymbol:B4GALT5
- Klonalitet:Monoclonal
- Klon:7E6
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Mouse
- Reaktivitet:Human
- Western blot:Yes
- Korsad adsorption:No
- Gen-ID:9334
- Antigen synonymer:gt-V|beta4Gal-T5|B4Gal-T5|BETA4-GALT-IV|beta4GalT-V
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:200 µl
- Förp.:200 µl
Type: Primary
Antigen: B4GALT5
Clonality: Monoclonal
Clone: 7E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human