Anti-NAE1 Mouse Polyclonal Antibody
ABNOH00008883-B01P
: Abnova
- Antibody type:Primary
- Antigen name:NEDD8 activating enzyme E1 subunit 1
- Antigen symbol:NAE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8883
- Antigen synonyms:APPBP1|A-116A10.1|ula-1|HPP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: NAE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat