Anti-DYRK4 Mouse Monoclonal Antibody [clone: 3B9]
ABNOH00008798-M04
: Abnova
- Antibody type:Primary
- Antigen name:Dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4
- Antigen symbol:DYRK4
- Clonality:Monoclonal
- Clone:3B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8798
- Antigen synonyms:Dyrk4b|AW049118|Dyrk4a
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: DYRK4
Clonality: Monoclonal
Clone: 3B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human