Anti-NOL4 Mouse Monoclonal Antibody [clone: 2A10]
ABNOH00008715-M01
: Abnova
- Antibody type:Primary
- Antigen name:Nucleolar protein 4
- Antigen symbol:NOL4
- Clonality:Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8715
- Antigen synonyms:CT125|HRIHFB2255|NOLP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NOL4
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human