Anti-PDE3A Mouse Monoclonal Antibody [clone: 5C12]
Catalog # ABNOH00005139-M01
Leverantör: Abnova
Specifikationer
- Antibody type:Primary
- Antigen name:Phosphodiesterase 3A, cGMP-inhibited
- Antigen symbol:PDE3A
- Clonality:Monoclonal
- Clone:5C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5139
- Antigen synonyms:CGI-PDE-A|HTNB|CGI-PDE|CGI-PDE A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifikationer
Om denna produkt
Type: Primary
Antigen: PDE3A
Clonality: Monoclonal
Clone: 5C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human