Anti-NPY Mouse Monoclonal Antibody [clone: 3F8]
ABNOH00004852-M04
: Abnova
- Antibody type:Primary
- Antigen name:Neuropeptide Y
- Antigen symbol:NPY
- Clonality:Monoclonal
- Clone:3F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4852
- Antigen synonyms:NPY|PYY4|OTTHUMP00000201947
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NPY
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human