Anti-HMGB1 Mouse Monoclonal Antibody [clone: 1E6-E10]
Catalog # ABNOH00003146-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:High-mobility Group Box 1
- Antigen symbol:HMGB1
- Clonality:Monoclonal
- Clone:1E6-E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3146
- Antigen synonyms:high-mobility group box 1|RP11-550P23.1|high mobility group protein 1|Amphoterin|SBP-1|high-mobility group (nonhistone chromosomal) protein 1|high mobility group protein B1|HMG3|Sulfoglucuronyl carbohydrate binding protein|HMG1|HMG-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HMGB1
Clonality: Monoclonal
Clone: 1E6-E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human