Anti-EWSR1 Mouse Monoclonal Antibody [clone: 5C10]
Catalog # ABNOH00002130-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:EWS RNA-binding protein 1
- Antigen symbol:EWSR1
- Clonality:Monoclonal
- Clone:5C10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2130
- Antigen synonyms:EWS-FLI1|EWS|bK984G1.4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: EWSR1
Clonality: Monoclonal
Clone: 5C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human