Anti-ADGRE1 Mouse Polyclonal Antibody
Catalog # ABNOH00002015-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Adhesion G Protein-Coupled Receptor E1
- Antigen symbol:ADGRE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2015
- Antigen synonyms:Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: ADGRE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human