Anti-EGR1 Mouse Monoclonal Antibody [clone: 6E8]
ABNOH00001958-M03
: Abnova
- Antibody type:Primary
- Antigen name:Early growth response protein 1
- Antigen symbol:EGR1
- Clonality:Monoclonal
- Clone:6E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1958
- Antigen synonyms:nerve growth factor-induced protein A|TIS8|AT225|ZNF225|ZIF-268|NGFI-A|G0S30|KROX-24
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: EGR1
Clonality: Monoclonal
Clone: 6E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human